missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Carboxy-5-fluorobenzeneboronic acid, 98%

Product Code. 15459667
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459667 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15459667 Supplier Alfa Aesar™ Supplier No. H52886.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 871329-84-9
Molecular Formula C7H6BFO4
Molecular Weight (g/mol) 183.929
MDL Number MFCD07363791
InChI Key APHPXZJIUTVQNC-UHFFFAOYSA-N
PubChem CID 44717205
IUPAC Name 3-borono-5-fluorobenzoic acid
SMILES B(C1=CC(=CC(=C1)F)C(=O)O)(O)O

Specifications

Melting Point 264°C to 266°C
Quantity 250 mg
Synonym 3-carboxy-5-fluorophenylboronic acid, 3-carboxy-5-fluoro benzeneboronic acid, 3-dihydroxyboranyl-5-fluorobenzoic acid, acmc-209qgr, 3-borono-5-fluorobenzoicacid, ksc657s3f, 3-borono-5-fluoro-benzoic acid, benzoic acid,3-borono-5-fluoro, benzoic acid, 3-borono-5-fluoro, 3-carboxy-5-fluorobenzeneboronic acid
Formula Weight 183.93
Percent Purity 98%
Chemical Name or Material 3-Carboxy-5-fluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.