CAS RN 874219-26-8
CAS RN 874219-26-8
IUPAC Name:
[3-(tert-butylcarbamoyl)-4-fluorophenyl]boronic acid
Synonyms:
3-(tert-butylcarbamoyl)-4-fluorophenylboronic acid
Molecular Weight (g/mol):
239.05
Molecular Formula:
C11H15BFNO3
InChi Key:
KHTWLZNHMILCQS-UHFFFAOYSA-N
SMILES:
CC(C)(C)NC(=O)C1=C(F)C=CC(=C1)B(O)O
1
–
1
of
1
results
1
Alfa Aesar™ 3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid, 98%
CAS: 874219-26-8 Molecular Formula: C11H15BFNO3 Molecular Weight (g/mol): 239.05 MDL Number: MFCD08235045 InChI Key: KHTWLZNHMILCQS-UHFFFAOYSA-N Synonym: 3-tert-butylcarbamoyl-4-fluorophenylboronic acid,3-tert-butylcarbamoyl-4-fluorophenyl boronic acid,3-tert-butylcarbamoyl-4-fluorobenzeneboronic acid,acmc-209qkr,3-tert-butylcarbamoyl-4-fluorophenyl boronicacid,3-tert-butylcarbamoyl-4-fluoranyl-phenyl boronic acid,3-tert-butylamino-oxomethyl-4-fluorophenyl boronic acid,boronic acid,b-3-1,1-dimethylethyl amino carbonyl-4-fluorophenyl PubChem CID: 44717587 IUPAC Name: [3-(tert-butylcarbamoyl)-4-fluorophenyl]boronic acid SMILES: CC(C)(C)NC(=O)C1=C(F)C=CC(=C1)B(O)O