missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid, 98%

Product Code. 15479925
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479925 250 mg 250mg
15469925 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15479925 Supplier Alfa Aesar™ Supplier No. H52594.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 874219-26-8
Molecular Formula C11H15BFNO3
Molecular Weight (g/mol) 239.05
MDL Number MFCD08235045
InChI Key KHTWLZNHMILCQS-UHFFFAOYSA-N
Synonym 3-tert-butylcarbamoyl-4-fluorophenylboronic acid, 3-tert-butylcarbamoyl-4-fluorophenyl boronic acid, 3-tert-butylcarbamoyl-4-fluorobenzeneboronic acid, acmc-209qkr, 3-tert-butylcarbamoyl-4-fluorophenyl boronicacid, 3-tert-butylcarbamoyl-4-fluoranyl-phenyl boronic acid, 3-tert-butylamino-oxomethyl-4-fluorophenyl boronic acid, boronic acid,b-3-1,1-dimethylethyl amino carbonyl-4-fluorophenyl
PubChem CID 44717587
IUPAC Name [3-(tert-butylcarbamoyl)-4-fluorophenyl]boronic acid
SMILES CC(C)(C)NC(=O)C1=C(F)C=CC(=C1)B(O)O

Specifications

Melting Point 218°C to 220°C
Quantity 250 mg
Formula Weight 239.05
Percent Purity 98%
Chemical Name or Material 3-tert-Butylcarbamoyl-4-fluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.