accessibility menu, dialog, popup

UserName

CAS RN 874288-39-8

BONH2FHOOH

CAS RN 874288-39-8

IUPAC Name: (4-carbamoyl-3-fluorophenyl)boronic acid
Synonyms: 4-carbamoyl-3-fluorophenylboronic acid
Molecular Weight (g/mol): 182.95
Molecular Formula: C7H7BFNO3
InChi Key: LKXXEWDYAWOZRW-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(F)C=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results