accessibility menu, dialog, popup

UserName

CAS RN 874289-51-7

BOHNH3CCH3H3CFHOOH

CAS RN 874289-51-7

IUPAC Name: [5-(tert-butylcarbamoyl)-2-fluorophenyl]boronic acid
Synonyms: 5-(tert-butylcarbamoyl)-2-fluorophenylboronic acid
Molecular Weight (g/mol): 239.05
Molecular Formula: C11H15BFNO3
InChi Key: KKSGPXWRHLBCCW-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results