missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15450356
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450356 250 mg 250mg
15440356 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15450356 Supplier Thermo Scientific Alfa Aesar Supplier No. H53080.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874289-51-7
Molecular Formula C11H15BFNO3
Molecular Weight (g/mol) 239.053
MDL Number MFCD08436054
InChI Key KKSGPXWRHLBCCW-UHFFFAOYSA-N
Synonym 5-tert-butylcarbamoyl-2-fluorophenyl boronic acid, n-t-butyl 3-borono-4-fluorobenzamide, 5-tert-butylcarbamoyl-2-fluorobenzeneboronic acid, n-tert-butyl 3-borono-4-fluorobenzamide, 5-tert-butylcarbamoyl-2-fluorophenylboronic acid, acmc-209qms, 5-tert-butylcarbamoyl-2-fluorophenyl boronicacid, 5-tert-butylcarbamoyl-2-fluoranyl-phenyl boronic acid, 5-tert-butylamino-oxomethyl-2-fluorophenyl boronic acid
PubChem CID 44717479
IUPAC Name [5-(tert-butylcarbamoyl)-2-fluorophenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NC(C)(C)C)F)(O)O

Specifications

Melting Point 142°C to 144°C
Quantity 250 mg
Formula Weight 239.05
Percent Purity 97%
Chemical Name or Material 5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.