accessibility menu, dialog, popup

UserName

CAS RN 88150-42-9

IUPAC Name: 3-ethyl 5-methyl 2-[(2-aminoethoxy)methyl]-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: amlodipine pelmec amlodipine, (+-)- (r)-isomer amlodipine norvasc pfizer brand of amlodipine besilate amlodipine besilate (substance) amlodipine besylate preparation amlodipine, (+-)-isomer amlodipine besylate (product)
Molecular Weight (g/mol): 408.88
Molecular Formula: C20H25ClN2O5
InChi Key: HTIQEAQVCYTUBX-UHFFFAOYNA-N
SMILES: CCOC(=O)C1=C(COCCN)NC(C)=C(C1C1=CC=CC=C1Cl)C(=O)OC
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)