accessibility menu, dialog, popup

UserName

HOONOO(E)

CAS RN 882-06-4

IUPAC Name: (2E)-3-(4-nitrophenyl)prop-2-enoic acid
Synonyms: trans-P-nitrocinnamic acid
Molecular Weight (g/mol): 193.16
Molecular Formula: C9H7NO4
InChi Key: XMMRNCHTDONGRJ-ZZXKWVIFSA-N
SMILES: OC(=O)\C=C\C1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results