missing translation for 'onlineSavingsMsg'
Learn More

trans-4-Nitrocinnamic acid, 98+%

Product Code. 11499863
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499863 100 g 100g
11489863 25 g 25g
11409873 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499863 Supplier Thermo Scientific Alfa Aesar Supplier No. A12990.22
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 882-06-4 | C9H7NO4 | 193.158 g/mol

trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-4-Nitrocinnamic acid, is used as a raw material for saccharine. It is also used as a useful chemical for biological applications. Some other applications include it is used as an organic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-06-4
Molecular Formula C9H7NO4
Molecular Weight (g/mol) 193.158
MDL Number MFCD00007381
InChI Key XMMRNCHTDONGRJ-ZZXKWVIFSA-N
Synonym 4-nitrocinnamic acid, 3-4-nitrophenyl acrylic acid, p-nitrocinnamic acid, cinnamic acid, p-nitro, 2-propenoic acid, 3-4-nitrophenyl, e-3-4-nitrophenyl acrylic acid, 3-4-nitrophenyl propenoic acid, trans-4-nitrocinnamic acid, ccris 1665, 2e-3-4-nitrophenyl prop-2-enoic acid
PubChem CID 737157
SMILES C1=CC(=CC=C1C=CC(=O)O)[N+](=O)[O-]

Specifications

Melting Point ∼290°C (decomposition)
Odor Odorless
Quantity 100 g
Beilstein 1369659
Solubility Information Insoluble in water.
Formula Weight 193.16
Percent Purity ≥98%
Chemical Name or Material trans-4-Nitrocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.