CAS RN 885269-91-0
IUPAC Name:
3-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}benzoic acid
Synonyms:
3-{4'-methoxy-[1,1'-biphenyl]-4-sulfonamido}benzoic acid
Molecular Weight (g/mol):
383.42
Molecular Formula:
C20H17NO5S
InChi Key:
ZBKATECYZDJGSI-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1)C1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC(=C1)C(O)=O
1
–
1
of
1
results
1
3-[4-(4-Methoxyphenyl)phenylsulfonamido]benzoic acid, Thermo Scientific™
CAS: 885269-91-0 Molecular Formula: C20H17NO5S Molecular Weight (g/mol): 383.418 MDL Number: MFCD06409494 InChI Key: ZBKATECYZDJGSI-UHFFFAOYSA-N Synonym: 3-4'-methoxy-1,1'-biphenyl-4-sulfonamido benzoic acid,3-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido benzoic acid,benzoicacid, 3-4'-methoxy 1,1'-biphenyl-4-yl sulfonyl amino,3-4'-methoxy 1,1'-biphenyl-4-sulfonyl amino benzoic acid PubChem CID: 44669193 IUPAC Name: 3-[[4-(4-methoxyphenyl)phenyl]sulfonylamino]benzoic acid SMILES: COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NC3=CC=CC(=C3)C(=O)O