missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(4-Methoxyphenyl)phenylsulfonamido]benzoic acid, Thermo Scientific™

Product Code. 15418135
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418135 250 mg 250mg
15408135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418135 Supplier Thermo Scientific Alfa Aesar Supplier No. H32583.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885269-91-0
Molecular Formula C20H17NO5S
Molecular Weight (g/mol) 383.418
MDL Number MFCD06409494
InChI Key ZBKATECYZDJGSI-UHFFFAOYSA-N
Synonym 3-4'-methoxy-1,1'-biphenyl-4-sulfonamido benzoic acid, 3-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido benzoic acid, benzoicacid, 3-4'-methoxy 1,1'-biphenyl-4-yl sulfonyl amino, 3-4'-methoxy 1,1'-biphenyl-4-sulfonyl amino benzoic acid
PubChem CID 44669193
IUPAC Name 3-[[4-(4-methoxyphenyl)phenyl]sulfonylamino]benzoic acid
SMILES COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NC3=CC=CC(=C3)C(=O)O

Spécification

Melting Point 235°C to 236°C
Quantity 250 mg
Formula Weight 383.43
Chemical Name or Material 3-[4-(4-Methoxyphenyl)phenylsulfonamido]benzoic acid
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.