accessibility menu, dialog, popup

UserName

ClFFFBr

CAS RN 886496-75-9

IUPAC Name: 1-(bromomethyl)-4-chloro-2-(trifluoromethyl)benzene
Synonyms: 1-(bromomethyl)-4-chloro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 273.48
Molecular Formula: C8H5BrClF3
InChi Key: LFFCKOROEHFKQA-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(CBr)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results