CAS RN 886496-91-9
IUPAC Name:
1-(bromomethyl)-3-chloro-5-(trifluoromethyl)benzene
Synonyms:
1-(bromomethyl)-3-chloro-5-(trifluoromethyl)benzene
Molecular Weight (g/mol):
273.48
Molecular Formula:
C8H5BrClF3
InChi Key:
DWXPOGQMJSSFCZ-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(Cl)=CC(CBr)=C1
1
–
1
of
1
results
1
3-Chloro-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
CAS: 886496-91-9 Molecular Formula: C8H5BrClF3 Molecular Weight (g/mol): 273.48 MDL Number: MFCD04115849 InChI Key: DWXPOGQMJSSFCZ-UHFFFAOYSA-N Synonym: 1-bromomethyl-3-chloro-5-trifluoromethyl benzene,3-chloro-5-trifluoromethyl benzyl bromide,3-chloro-5-trifluoromethylbenzyl bromide,1-bromomethyl-3-chloro-5-trifluoromethyl-benzene,1-bromomethyl-3-chloro-5-trifluoromethyl benzen,1-bromomethyl-3-chloranyl-5-trifluoromethyl benzene PubChem CID: 3714672 IUPAC Name: 1-(bromomethyl)-3-chloro-5-(trifluoromethyl)benzene SMILES: FC(F)(F)C1=CC(Cl)=CC(CBr)=C1