missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11332669
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332669 5 g 5g
11322669 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332669 Supplier Thermo Scientific Alfa Aesar Supplier No. H26709.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
API. As pharmaceuticals, they find applications in antibiotics, as sedatives, and for cancer treatment. Used frequently in organic synthesis. Find application as propellants.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886496-91-9
Molecular Formula C8H5BrClF3
Molecular Weight (g/mol) 273.48
MDL Number MFCD04115849
InChI Key DWXPOGQMJSSFCZ-UHFFFAOYSA-N
Synonym 1-bromomethyl-3-chloro-5-trifluoromethyl benzene, 3-chloro-5-trifluoromethyl benzyl bromide, 3-chloro-5-trifluoromethylbenzyl bromide, 1-bromomethyl-3-chloro-5-trifluoromethyl-benzene, 1-bromomethyl-3-chloro-5-trifluoromethyl benzen, 1-bromomethyl-3-chloranyl-5-trifluoromethyl benzene
PubChem CID 3714672
IUPAC Name 1-(bromomethyl)-3-chloro-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Cl)=CC(CBr)=C1

Specifications

Quantity 5 g
UN Number UN3265
Formula Weight 273.48
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.