accessibility menu, dialog, popup

UserName

CAS RN 886498-08-4

IUPAC Name: 2-fluoro-5-(trifluoromethoxy)benzonitrile
Synonyms: 2-fluoro-5-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 205.11
Molecular Formula: C8H3F4NO
InChi Key: MFLGNKVMGUFLOV-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC(F)(F)F)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results