accessibility menu, dialog, popup

UserName

CH3OHHOO

CAS RN 89-56-5

IUPAC Name: 2-hydroxy-5-methylbenzoic acid
Synonyms: 5-methylsalicylic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: DLGBEGBHXSAQOC-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(=C1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)