missing translation for 'onlineSavingsMsg'
Learn More

5-Methylsalicylic acid, 98%

Product Code. 10455125 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10455125 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10455125 Supplier Thermo Scientific Acros Supplier No. 163860500
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 89-56-5 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 89-56-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 151°C to 155°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C8H8O3
Linear Formula CH3C6H3(OH)CO2H
MDL Number MFCD00002461
Quantity 50 g
Beilstein 10, 227
Merck Index 15, 2571
Synonym 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid
InChI Key DLGBEGBHXSAQOC-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)O)C(=O)O
IUPAC Name 2-hydroxy-5-methylbenzoic acid
Molecular Weight (g/mol) 152.15
PubChem CID 6973
Formula Weight 152.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Methylsalicylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methylsalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.