CAS RN 892502-29-3
IUPAC Name:
{2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol
Synonyms:
{2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]phenyl}methanol
Molecular Weight (g/mol):
256.23
Molecular Formula:
C12H11F3N2O
InChi Key:
HLMRFDCLSUXVQC-UHFFFAOYSA-N
SMILES:
CN1N=C(C=C1C1=CC=CC=C1CO)C(F)(F)F
1
–
1
of
1
results
1
{2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol, 97%, Thermo Scientific™
CAS: 892502-29-3 Molecular Formula: C12H11F3N2O Molecular Weight (g/mol): 256.228 MDL Number: MFCD09817511 InChI Key: HLMRFDCLSUXVQC-UHFFFAOYSA-N Synonym: 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl phenyl methanol,2-2-methyl-5-trifluoromethyl pyrazol-3-yl phenyl methanol,2-1-methyl-3-trifluoromethyl pyrazol-5-yl phenyl methan-1-ol,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzyl alcohol,benzenemethanol,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl PubChem CID: 24229659 IUPAC Name: [2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]phenyl]methanol SMILES: CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2CO