missing translation for 'onlineSavingsMsg'
Learn More

{2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol, 97%, Thermo Scientific™

Product Code. 10411894
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10411894 250 mg Amber Glass Bottle 250mg
10021974 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10411894 Supplier Thermo Scientific Maybridge Supplier No. CC62109CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-29-3
Molecular Formula C12H11F3N2O
Molecular Weight (g/mol) 256.228
MDL Number MFCD09817511
InChI Key HLMRFDCLSUXVQC-UHFFFAOYSA-N
Synonym 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl phenyl methanol, 2-2-methyl-5-trifluoromethyl pyrazol-3-yl phenyl methanol, 2-1-methyl-3-trifluoromethyl pyrazol-5-yl phenyl methan-1-ol, 2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl benzyl alcohol, benzenemethanol,2-1-methyl-3-trifluoromethyl-1h-pyrazol-5-yl
PubChem CID 24229659
IUPAC Name [2-[2-methyl-5-(trifluoromethyl)pyrazol-3-yl]phenyl]methanol
SMILES CN1C(=CC(=N1)C(F)(F)F)C2=CC=CC=C2CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 256.23
Percent Purity 97%
Chemical Name or Material {2-[1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]phenyl}methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.