accessibility menu, dialog, popup

UserName

CAS RN 90-64-2

IUPAC Name: 2-hydroxy-2-phenylacetic acid
Synonyms: mandelic acid 扁桃酸
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: IWYDHOAUDWTVEP-UHFFFAOYNA-N
SMILES: OC(C(O)=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (7)
  • (5)
Percent Purity 
  • (4)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (7)
  • (5)
  • (4)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)