accessibility menu, dialog, popup

UserName

OONHOOHOH

CAS RN 900799-69-1

IUPAC Name: 3-{[(benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid
Synonyms: 3-{[(benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid
Molecular Weight (g/mol): 287.27
Molecular Formula: C15H13NO5
InChi Key: DIPYTSQSLOJNFI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(NC(=O)OCC2=CC=CC=C2)=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results