missing translation for 'onlineSavingsMsg'
Learn More

3-{[(Benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid, 97%, Thermo Scientific™

Product Code. 11306905
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11306905 5 g Amber Glass Bottle 5g
10401694 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11306905 Supplier Thermo Scientific Maybridge Supplier No. MO08033DE
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 900799-69-1
Molecular Formula C15H13NO5
Molecular Weight (g/mol) 287.271
MDL Number MFCD09817564
InChI Key DIPYTSQSLOJNFI-UHFFFAOYSA-N
Synonym 3-cbz-amino-5-hydroxybenzoic acid, 3-benzyloxy carbonyl amino-5-hydroxybenzoic acid, 3-n-cbz-amino-5-hydroxybenzoic acid, 3-benzyloxycarbonylamino-5-hydroxy-benzoic acid, benzoicacid, 3-hydroxy-5-phenylmethoxy carbonyl amino, benzoicacid,3-hydroxy-5-phenylmethoxy carbonyl amino
PubChem CID 24229785
IUPAC Name 3-hydroxy-5-(phenylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC2=CC(=CC(=C2)C(=O)O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 287.27
Percent Purity 97%
Chemical Name or Material 3-{[(Benzyloxy)carbonyl]amino}-5-hydroxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.