CAS RN 906352-62-3
IUPAC Name:
2-[(1H-1,2,4-triazol-1-yl)methyl]benzaldehyde
Synonyms:
2-(1,2,4-triazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol):
187.2
Molecular Formula:
C10H9N3O
InChi Key:
XPXZXSFDTOZFQQ-UHFFFAOYSA-N
SMILES:
O=CC1=CC=CC=C1CN1C=NC=N1
1
–
1
of
1
results
1
2-(1H-1,2,4-triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™
CAS: 906352-62-3 Molecular Formula: C10H9N3O Molecular Weight (g/mol): 187.202 MDL Number: MFCD09025843 InChI Key: XPXZXSFDTOZFQQ-UHFFFAOYSA-N Synonym: 2-1h-1,2,4-triazol-1-ylmethyl benzaldehyde,2-1,2,4-triazol-1-ylmethyl benzaldehyde,2-1h-1,2,4-triazol-1-yl methyl benzaldehyde,2-1,2,4-triazolylmethyl benzaldehyde PubChem CID: 18525815 IUPAC Name: 2-(1,2,4-triazol-1-ylmethyl)benzaldehyde SMILES: C1=CC=C(C(=C1)CN2C=NC=N2)C=O