missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-triazol-1-ylmethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 11330657
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11330657 250 mg Amber Glass Bottle 250mg
10788065 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330657 Supplier Thermo Scientific Maybridge Supplier No. CC47504CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-62-3
Molecular Formula C10H9N3O
Molecular Weight (g/mol) 187.202
MDL Number MFCD09025843
InChI Key XPXZXSFDTOZFQQ-UHFFFAOYSA-N
Synonym 2-1h-1,2,4-triazol-1-ylmethyl benzaldehyde, 2-1,2,4-triazol-1-ylmethyl benzaldehyde, 2-1h-1,2,4-triazol-1-yl methyl benzaldehyde, 2-1,2,4-triazolylmethyl benzaldehyde
PubChem CID 18525815
IUPAC Name 2-(1,2,4-triazol-1-ylmethyl)benzaldehyde
SMILES C1=CC=C(C(=C1)CN2C=NC=N2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 2-(1H-1, 2, 4-Triazol-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.