accessibility menu, dialog, popup

UserName

OOI

CAS RN 906352-70-3

IUPAC Name: 4-(2-iodophenoxy)oxane
Synonyms: 4-(2-iodophenoxy)oxane
Molecular Weight (g/mol): 304.13
Molecular Formula: C11H13IO2
InChi Key: ADTBIBCNKWWYRD-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1OC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results