accessibility menu, dialog, popup

UserName

CAS RN 91-53-2

H3COHNCH3CH3CH3

CAS RN 91-53-2

IUPAC Name: 6-ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Synonyms: ethoxyquin
Molecular Weight (g/mol): 217.31
Molecular Formula: C14H19NO
InChi Key: DECIPOUIJURFOJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2NC(C)(C)C=C(C)C2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)
Boiling Point 
  • (2)
Concentration 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)