accessibility menu, dialog, popup

UserName

CH3NNCl

CAS RN 912569-61-0

IUPAC Name: 3-[3-(chloromethyl)phenyl]-1-methyl-1H-pyrazole
Synonyms: 3-[3-(chloromethyl)phenyl]-1-methylpyrazole
Molecular Weight (g/mol): 206.67
Molecular Formula: C11H11ClN2
InChi Key: XEIKNZPKJWESHW-UHFFFAOYSA-N
SMILES: CN1C=CC(=N1)C1=CC(CCl)=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results