missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(chloromethyl)phenyl]-1-methyl-1H-pyrazole, Tech., Thermo Scientific™

Product Code. 10327694
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10327694 1 g Amber Glass Bottle 1g
10225674 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10327694 Supplier Thermo Scientific Maybridge Supplier No. CC60624DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 912569-61-0
Molecular Formula C11H11ClN2
Molecular Weight (g/mol) 206.673
MDL Number MFCD09702402
InChI Key XEIKNZPKJWESHW-UHFFFAOYSA-N
Synonym 3-3-chloromethyl phenyl-1-methyl-1h-pyrazole, 3-3-chloromethyl phenyl-1-methylpyrazole
PubChem CID 24229632
IUPAC Name 3-[3-(chloromethyl)phenyl]-1-methylpyrazole
SMILES CN1C=CC(=N1)C2=CC(=CC=C2)CCl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.67
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-[3-(chloromethyl)phenyl]-1-methyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.