accessibility menu, dialog, popup

UserName

BrF

CAS RN 91319-54-9

IUPAC Name: 1-(2-bromoethyl)-2-fluorobenzene
Synonyms: 1-(2-bromoethyl)-2-fluorobenzene
Molecular Weight (g/mol): 203.05
Molecular Formula: C8H8BrF
InChi Key: FQGDFWIQBCQXPS-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CCBr
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results