missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenethyl bromide, 97%

Product Code. 11914541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11914541 5 g 5g
11904541 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11914541 Supplier Thermo Scientific Acros Supplier No. 440120050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91319-54-9 | C8H8BrF | 203.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91319-54-9
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
InChI Key FQGDFWIQBCQXPS-UHFFFAOYSA-N
Synonym 2-fluorophenethyl bromide, 1-2-bromoethyl-2-fluorobenzene, 1-2-bromo-ethyl-2-fluoro-benzene, benzene, 1-2-bromoethyl-2-fluoro, 2-2-bromoethyl-1-fluorobenzene, 1-2-bromoethyl-2-fluoro-benzene, 2-fluorophenyl-ethylbromide, acmc-209wk4, 2-fluorophenyl ethyl bromide, 2-2-fluorophenyl-ethylbromide
PubChem CID 2756964
IUPAC Name 1-(2-bromoethyl)-2-fluorobenzene
SMILES C1=CC=C(C(=C1)CCBr)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 73°C to 75°C (20.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 203.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenethyl bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluorophenethyl bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.