accessibility menu, dialog, popup

UserName

CAS RN 92-48-8

H3COO

CAS RN 92-48-8

IUPAC Name: 6-methyl-2H-chromen-2-one
Synonyms: 6-methylcoumarin
Molecular Weight (g/mol): 160.17
Molecular Formula: C10H8O2
InChi Key: FXFYOPQLGGEACP-UHFFFAOYSA-N
SMILES: CC1=CC=C2OC(=O)C=CC2=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results