missing translation for 'onlineSavingsMsg'
Learn More

6-Methylcoumarin, 99%

Product Code. 11453977
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453977 50 g 50g
11384639 100 g 100g
11463977 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11453977 Supplier Thermo Scientific Alfa Aesar Supplier No. A19382.18
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-48-8 | C10H8O2 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 92-48-8
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00006875
InChI Key FXFYOPQLGGEACP-UHFFFAOYSA-N
Synonym 6-methylcoumarin, 6-methylbenzopyrone, 6-methyl-2h-chromen-2-one, toncarine, 6-methyl coumarin, methyl coumarin, 6-methylcoumarinic anhydride, coumarin, 6-methyl, 6-methyl-1,2-benzopyrone, 6-methylcumarin
PubChem CID 7092
ChEBI CHEBI:563586
IUPAC Name 6-methylchromen-2-one
SMILES CC1=CC=C2OC(=O)C=CC2=C1

Tekniske data

Melting Point 74°C to 77°C
Boiling Point 303°C (725 mmHg)
Quantity 50 g
Beilstein 4222
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 6-Methylcoumarin

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.