accessibility menu, dialog, popup

UserName

CAS RN 92-51-3

IUPAC Name: 1,1'-bi(cyclohexane)
Synonyms: dicyclohexyl
Molecular Weight (g/mol): 166.31
Molecular Formula: C12H22
InChi Key: WVIIMZNLDWSIRH-UHFFFAOYSA-N
SMILES: C1CCC(CC1)C1CCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)