missing translation for 'onlineSavingsMsg'
Learn More

Bicyclohexyl, 98%

Product Code. 11455563
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455563 50 g 50g
11445563 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455563 Supplier Thermo Scientific Alfa Aesar Supplier No. A13481.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-51-3 | C12H22 | 166.31 g/mol

Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bicyclohexyl is used as process media or solvent in many industries including chemicals, petrochemicals, polymers, coatings, photography. And it is also used to produce 1-fluorobicyclohexyl at temperature of -75°C.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-51-3
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003815
InChI Key WVIIMZNLDWSIRH-UHFFFAOYSA-N
Synonym bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro
PubChem CID 7094
IUPAC Name cyclohexylcyclohexane
SMILES C1CCC(CC1)C1CCCCC1

Specifications

Melting Point 3°C to 4°C
Density 0.864
Boiling Point 226°C to 228°C
Flash Point 92°C (198°F)
Refractive Index 1.478
Quantity 50 g
Beilstein 1848266
Solubility Information Difficult to mix.
Formula Weight 166.31
Percent Purity 98%
Chemical Name or Material Bicyclohexyl
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.