missing translation for 'onlineSavingsMsg'
Learn More

Bicyclohexyl, 99%

Product Code. 10502014 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10502014 25 g Glass bottle 25g
10361391 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10502014 Supplier Thermo Scientific Acros Supplier No. 402210250
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-51-3 | C12H22 | 166.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-51-3
Molecular Formula C12H22
Molecular Weight (g/mol) 166.31
MDL Number MFCD00003815
InChI Key WVIIMZNLDWSIRH-UHFFFAOYSA-N
Synonym bicyclohexyl, 1,1'-bicyclohexyl, dicyclohexyl, bicyclohexane, dicyclohexane, dodecahydrobiphenyl, cyclohexane, cyclohexyl, 1,1'-biphenyl, dodecahydro, 1,1'-bi cyclohexyl, 1, dodecahydro
PubChem CID 7094
IUPAC Name cyclohexylcyclohexane
SMILES C1CCC(CC1)C1CCCCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 3.0°C to 4.0°C
Color Colorless
Density 0.8600g/mL
Boiling Point 227.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4780 to 1.4800
Quantity 25 g
Beilstein 05, 108
Specific Gravity 0.86
Solubility Information Solubility in water: slightly soluble.
Formula Weight 166.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Bicyclohexyl, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.