accessibility menu, dialog, popup

UserName

CAS RN 92-70-6

IUPAC Name: 3-hydroxynaphthalene-2-carboxylic acid
Synonyms: BONA bon acid miketazol developer ons C.I. developer 20 (obs.) kyselina 3-hydroxy-2-naftoova BON C.I. developer 20 naphthol B.O.N. CI developer 20 3-hydroxy-2-naphthoesaeure
Molecular Weight (g/mol): 188.18
Molecular Formula: C11H8O3
InChi Key: ALKYHXVLJMQRLQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)