missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic acid, 98%

Product Code. 11489486
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489486 250 g 250g
11479486 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489486 Supplier Thermo Scientific Alfa Aesar Supplier No. A19214.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-70-6 | C11H8O3 | 188.182 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-70-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00004103
InChI Key ALKYHXVLJMQRLQ-UHFFFAOYSA-N
Synonym 3-hydroxy-2-naphthoic acid, 2-hydroxy-3-naphthoic acid, bon acid, bona, developer bon, 2-naphthol-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy, b.o.n. acid, c.i. developer 20, miketazol developer ons
PubChem CID 7104
ChEBI CHEBI:80383
IUPAC Name 3-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O

Specifications

Melting Point 218°C to 222°C
Flash Point 220°C (428°F)
Quantity 250 g
Beilstein 744100
Merck Index 14,4835
Formula Weight 188.18
Percent Purity 98%
Chemical Name or Material 3-Hydroxy-2-naphthoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.