CAS RN 922510-21-2
CAS RN 922510-21-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Phenanthrol-d9, TRC
CAS: 922510-21-2 Molecular Formula: C14HD9O Molecular Weight (g/mol): 203.28 Synonym: Phenanthren-1,2,3,5,6,7,8,9,10-d9-4-ol,4-Phenanthrenol-d9,4-Phenanthrol-d9,4-Hydroxyphenanthrene-d9,NSC 171284-d9 IUPAC Name: phenanthren-d9-4-ol SMILES: [2H]C1=C([2H])C([2H])=C([2H])C2=C1C([2H])=C([2H])C3=C([2H])C([2H])=C([2H])C(O)=C32