accessibility menu, dialog, popup

UserName

CAS RN 92439-20-8

OOOHOH

CAS RN 92439-20-8

IUPAC Name: 6-hydroxy-2-(2-hydroxyphenyl)-4H-chromen-4-one
Synonyms: 6-hydroxy-2-(2-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: YCGXYGWBHFKQHY-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC(=CC(=O)C2=C1)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)