accessibility menu, dialog, popup

UserName

H3COClClClHOO

CAS RN 93-72-1

IUPAC Name: 2-(2,4,5-trichlorophenoxy)propanoic acid
Synonyms: double strength C9H7cl3O3 fenoprop, (S)-isomer caswell no. 739 fenoprop, (R)-isomer color-set fenormone amchem 2,4,5-TP herbicides, silvex (R)-fenoprop
Molecular Weight (g/mol): 269.5
Molecular Formula: C9H7Cl3O3
InChi Key: ZLSWBLPERHFHIS-UHFFFAOYNA-N
SMILES: CC(OC1=CC(Cl)=C(Cl)C=C1Cl)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results