accessibility menu, dialog, popup

UserName

CAS RN 931-86-2

NH2NNOHN

CAS RN 931-86-2

IUPAC Name: 6-amino-1,2-dihydro-1,3,5-triazin-2-one
Synonyms: 4-amino-3H-1,3,5-triazin-2-one
Molecular Weight (g/mol): 112.09
Molecular Formula: C3H4N4O
InChi Key: MFEFTTYGMZOIKO-UHFFFAOYSA-N
SMILES: NC1=NC=NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results