accessibility menu, dialog, popup

UserName

CH3NHSOONO

CAS RN 941717-08-4

IUPAC Name: methyl({[2-(morpholine-4-sulfonyl)phenyl]methyl})amine
Synonyms: methyl({[2-(morpholine-4-sulfonyl)phenyl]methyl})amine
Molecular Weight (g/mol): 270.35
Molecular Formula: C12H18N2O3S
InChi Key: ASGSHKVYZRSASF-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results