missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(morpholinosulfonyl)benzylamine, 97%, Thermo Scientific™

Product Code. 10412264
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10412264 1 g Amber Glass Bottle 1g
10163274 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10412264 Supplier Thermo Scientific Maybridge Supplier No. CC69646DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 941717-08-4
Molecular Formula C12H18N2O3S
Molecular Weight (g/mol) 270.35
MDL Number MFCD09879987
InChI Key ASGSHKVYZRSASF-UHFFFAOYSA-N
Synonym n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
PubChem CID 24229774
IUPAC Name N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine
SMILES CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 270.35
Percent Purity 97%
Chemical Name or Material N-methyl-2-(morpholinosulfonyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.