accessibility menu, dialog, popup

UserName

CAS RN 95-94-3

IUPAC Name: 1,2,4,5-tetrachlorobenzene
Synonyms: 1,2,4,5-tetrachlorobenzene s-tetrachlorobenzene ccris 766 rcra waste number U207
Molecular Weight (g/mol): 215.88
Molecular Formula: C6H2Cl4
InChi Key: JHBKHLUZVFWLAG-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)C=C1Cl
Molecular Weight (g/mol) 
  • (10)
  • (6)
Solvent or Matrix 
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Concentration 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (10)
  • (6)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)