missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 8270 App. IX Calibration Mixture 602 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30309947
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309947 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309947 Supplier Dr. Ehrenstorfer Supplier No. DREA50000602DI
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,4,5-Tetrachlorobenzene∣1,3,5-Trinitrobenzene∣1,3-Dinitrobenzene∣1-Aminonaphthalene∣2,3,4,6-Tetrachlorophenol∣2,6-Dichlorophenol∣2-Acetamidofluoren∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)∣2-Aminonaphthalene∣2-Picolin (2-Methylpyridine)∣3-Methylcholanthrene∣3-Methylphenol (m-Cresol)∣4-Aminobiphenyl∣4-Nitroquinoline-1-oxide∣4-Nitrosomorpholine∣7,12-Dimethylbenzo(a)anthracene∣Acetophenone∣Aniline∣Benzyl alcohol∣Dinoseb∣Diphenylamine∣Hexachloropropene (Perchloropropene)∣Isosafrole∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine∣o-Toluidine∣Pentachlorobenzene∣Pentachloroethane∣Phenacetin∣Pyridine∣Quintozene∣Safrole∣Sudan Yellow
CAS 95-94-3, 99-35-4, 99-65-0, 134-32-7, 58-90-2, 87-65-0
Analyte or Component Names 1,2,4,5-Tetrachlorobenzene∣1,3,5-Trinitrobenzene∣1,3-Dinitrobenzene∣1-Aminonaphthalene∣2,3,4,6-Tetrachlorophenol∣2,6-Dichlorophenol∣2-Acetamidofluoren∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline)∣2-Aminonaphthalene∣2-Picolin (2-Methylpyridine)∣3-Methylcholanthrene∣3-Methylphenol (m-Cresol)∣4-Aminobiphenyl∣4-Nitroquinoline-1-oxide∣4-Nitrosomorpholine∣7,12-Dimethylbenzo(a)anthracene∣Acetophenone∣Aniline∣Benzyl alcohol∣Dinoseb∣Diphenylamine∣Hexachloropropene (Perchloropropene)∣Isosafrole∣N-Nitroso-diethylamine∣N-Nitroso-di-n-butylamine∣N-Nitrosomethylethylamine∣N-Nitrosopiperidine∣N-Nitrosopyrrolidine∣o-Toluidine∣Pentachlorobenzene∣Pentachloroethane∣Phenacetin∣Pyridine∣Quintozene∣Safrole∣Sudan Yellow
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,2,4,5-Tetrachlorobenzene 1000 μg/mL∣1,3,5-Trinitrobenzene 1000 μg/mL∣1,3-Dinitrobenzene 1000 μg/mL∣1-Aminonaphthalene 1000 μg/mL∣2,3,4,6-Tetrachlorophenol 1000 μg/mL∣2,6-Dichlorophenol 1000 μg/mL∣2-Acetamidofluoren 1000 μg/mL∣2-Amino-4-nitrotoluene (2-Methyl-5-nitroaniline) 1000 μg/mL∣2-Aminonaphthalene 1000 μg/mL∣2-Picolin (2-Methylpyridine) 1000 μg/mL∣3-Methylcholanthrene 1000 μg/mL∣3-Methylphenol (m-Cresol) 1000 μg/mL∣4-Aminobiphenyl 1000 μg/mL∣4-Nitroquinoline-1-oxide 1000 μg/mL∣4-Nitrosomorpholine 1000 μg/mL∣7,12-Dimethylbenzo(a)anthracene 1000 μg/mL∣Acetophenone 1000 μg/mL∣Aniline 1000 μg/mL∣Benzyl alcohol 1000 μg/mL∣Dinoseb 1000 μg/mL∣Diphenylamine 1000 μg/mL∣Hexachloropropene (Perchloropropene) 1000 μg/mL∣Isosafrole 1000 μg/mL∣N-Nitroso-diethylamine 1000 μg/mL∣N-Nitroso-di-n-butylamine 1000 μg/mL∣N-Nitrosomethylethylamine 1000 μg/mL∣N-Nitrosopiperidine 1000 μg/mL∣N-Nitrosopyrrolidine 1000 μg/mL∣o-Toluidine 1000 μg/mL∣Pentachlorobenzene 1000 μg/mL∣Pentachloroethane 1000 μg/mL∣Phenacetin 1000 μg/mL∣Pyridine 1000 μg/mL∣Quintozene 1000 μg/mL∣Safrole 1000 μg/mL∣Sudan Yellow 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.