accessibility menu, dialog, popup

UserName

CH3(R)H3CH3N

CAS RN 96-15-1

IUPAC Name: (2R)-2-methylbutan-1-aminium
Synonyms: (2R)-2-methylbutan-1-aminium
Molecular Weight (g/mol): 88.17
Molecular Formula: C5H14N
InChi Key: VJROPLWGFCORRM-RXMQYKEDSA-O
SMILES: CC[C@@H](C)C[NH3+]
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)