missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutylamine, 95%, Thermo Scientific™

Product Code. 10452733
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10452733 10 g Amber Glass Bottle 10g
10125772 1 g Amber Glass Bottle 1g
10548744 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10452733 Supplier Thermo Scientific Maybridge Supplier No. SB01376EA
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 96-15-1
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.166
MDL Number MFCD00008147
InChI Key VJROPLWGFCORRM-UHFFFAOYSA-N
Synonym 2-methylbutylamine, 1-butanamine, 2-methyl, 1-amino-2-methylbutane, 2-methylbutyl amine, 2-methyl-1-butylamine, 2-methylbutanamine, 2-methyl-1-butanamine, butylamine, 2-methyl, 1-amino-2-methyl-butane, 2-methyl-butylamine
PubChem CID 7283
IUPAC Name 2-methylbutan-1-amine
SMILES CCC(C)CN

Specifications

CAS Min % 95.0
Density 0.7300g/mL
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula C2H5CH(CH3)CH2NH2
Quantity 10 g
Formula Weight 87.16
Percent Purity 95%
Chemical Name or Material 2-Methylbutylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.