accessibility menu, dialog, popup

UserName

CAS RN 96-24-2

OHHOCl

CAS RN 96-24-2

IUPAC Name: 3-chloropropane-1,2-diol
Synonyms: α chlorohydrin 3-mcpd monochlorohydrin α-chlorhydrin glycerin α-monochlorhydrin glyceryl α-chlorohydrin C3H7ClO2 glyceryl-α-chlorohydrin glycerin epichlorohydrin epibloc
Molecular Weight (g/mol): 110.54
Molecular Formula: C3H7ClO2
InChi Key: SSZWWUDQMAHNAQ-UHFFFAOYNA-N
SMILES: OCC(O)CCl
Molecular Weight (g/mol) 
  • (5)
  • (8)
Percent Purity 
  • (3)
  • (4)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (3)
  • (5)
  • (8)
  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)