accessibility menu, dialog, popup

UserName

CAS RN 96-50-4

IUPAC Name: 1,3-thiazol-2-amine
Synonyms: aminothiazole
Molecular Weight (g/mol): 100.14
Molecular Formula: C3H4N2S
InChi Key: RAIPHJJURHTUIC-UHFFFAOYSA-N
SMILES: NC1=NC=CS1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (8)
  • (2)
Boiling Point 
  • (5)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (8)
  • (2)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)