accessibility menu, dialog, popup

UserName

CAS RN 96-99-1

HOOClNOO

CAS RN 96-99-1

IUPAC Name: 4-chloro-3-nitrobenzoic acid
Synonyms: 4-chloro-3-nitrobenzoic acid
Molecular Weight (g/mol): 201.56
Molecular Formula: C7H4ClNO4
InChi Key: DFXQXFGFOLXAPO-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results